Anti-MASTL Rabbit Polyclonal Antibody

Lieferant: Abgent
AP7147D
ABGEAP7147DEA 354 EUR
ABGEAP7147D
Anti-MASTL Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: MASTL (microtubule associated serine/threonine kinase-like)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR