Anti-MELK Rabbit Polyclonal Antibody

Lieferant: Abgent
AP7149B
ABGEAP7149BEA 354 EUR
ABGEAP7149B
Anti-MELK Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: MELK (maternal embryonic leucine zipper kinase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR