Anti-NME3 Rabbit Polyclonal Antibody

Lieferant: Abgent
AP7156A
ABGEAP7156AEA 354 EUR
ABGEAP7156A
Anti-NME3 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: NME3 (NME/NM23 nucleoside diphosphate kinase 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR