Anti-RICTOR Rabbit Polyclonal Antibody
Lieferant: Abgent
AP7259C
ABGEAP7259CEA 354 EUR
ABGEAP7259C
Anti-RICTOR Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: RICTOR (RPTOR independent companion of MTOR, complex 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse