Anti-MME Rabbit Polyclonal Antibody

Lieferant: Abgent
AP7329C
ABGEAP7329CEA 354 EUR
ABGEAP7329C
Anti-MME Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: MME (membrane metallo-endopeptidase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR