Anti-DDX39A Rabbit Polyclonal Antibody

Lieferant: Abgent
AP7451D
ABGEAP7451DEA 354 EUR
ABGEAP7451D
Anti-DDX39A Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: DDX39A (DEAD (Asp-Glu-Ala-Asp) box polypeptide 39A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR