Anti-HNF4G Rabbit Polyclonal Antibody

Lieferant: Abgent
AP7452C
ABGEAP7452CEA 354 EUR
ABGEAP7452C
Anti-HNF4G Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: HNF4G (hepatocyte nuclear factor 4, gamma)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR