Anti-CDK2 Rabbit Polyclonal Antibody

Lieferant: Abgent
AP7518B
ABGEAP7518BEA 354 EUR
ABGEAP7518B
Anti-CDK2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CDK2 (cyclin-dependent kinase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR