Anti-DYRK2 Rabbit Polyclonal Antibody
Lieferant: Abgent
AP7534A
ABGEAP7534AEA 354 EUR
ABGEAP7534A
Anti-DYRK2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: DYRK2 (dual-specificity tyrosine-(Y)-phosphorylation regulated kinase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human