Anti-PDGFRA Rabbit Polyclonal Antibody
Lieferant: Abgent
AP7666H
ABGEAP7666HEA 354 EUR
ABGEAP7666H
Anti-PDGFRA Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PDGFRA (platelet-derived growth factor receptor, alpha polypeptide)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human