Anti-HCK Rabbit Polyclonal Antibody

Lieferant: Abgent
AP7710D
ABGEAP7710DEA 354 EUR
ABGEAP7710D
Anti-HCK Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: HCK (hemopoietic cell kinase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR