Anti-FKBP1B Rabbit Polyclonal Antibody

Lieferant: Abgent
AP7868A
ABGEAP7868AEA 354 EUR
ABGEAP7868A
Anti-FKBP1B Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: FKBP1B (FK506 binding protein 1B, 12.6 kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR