Anti-ALOX5AP Rabbit Polyclonal Antibody

Lieferant: Abgent
AP7897C
ABGEAP7897CEA 354 EUR
ABGEAP7897C
Anti-ALOX5AP Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ALOX5AP (arachidonate 5-lipoxygenase-activating protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR