Anti-NUAK1 Rabbit Polyclonal Antibody

Lieferant: Abgent
AP8066A
ABGEAP8066AEA 354 EUR
ABGEAP8066A
Anti-NUAK1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: NUAK1 (NUAK family, SNF1-like kinase, 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR