Anti-PPM1F Rabbit Polyclonal Antibody
Lieferant: Abgent
AP8459A
ABGEAP8459AEA 354 EUR
ABGEAP8459A
Anti-PPM1F Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PPM1F (protein phosphatase, Mg2+/Mn2+ dependent, 1F)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human