Anti-DUSP1 Rabbit Polyclonal Antibody

Lieferant: Abgent
AP8470A
ABGEAP8470AEA 354 EUR
ABGEAP8470A
Anti-DUSP1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: DUSP1 (dual specificity phosphatase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR