Anti-MICA Rabbit Polyclonal Antibody

Lieferant: Abgent
AP8626C
ABGEAP8626CEA 354 EUR
ABGEAP8626C
Anti-MICA Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: MICA (MHC class I polypeptide-related sequence A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR