Anti-PIN1 Rabbit Polyclonal Antibody

Lieferant: Abgent
AP8959C
ABGEAP8959CEA 354 EUR
ABGEAP8959C
Anti-PIN1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PIN1 (peptidylprolyl cis/trans isomerase, NIMA-interacting 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR