Anti-LY75 Rabbit Polyclonal Antibody

Lieferant: Abgent
AP8994C
ABGEAP8994CEA 354 EUR
ABGEAP8994C
Anti-LY75 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: LY75 (lymphocyte antigen 75)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR