Anti-CFLAR Rabbit Polyclonal Antibody

Lieferant: Abgent
AP9123C
ABGEAP9123CEA 354 EUR
ABGEAP9123C
Anti-CFLAR Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CFLAR (CASP8 and FADD-like apoptosis regulator)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR