Anti-TFPT Rabbit Polyclonal Antibody

Lieferant: Abgent
AP9304A
ABGEAP9304AEA 354 EUR
ABGEAP9304A
Anti-TFPT Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TFPT (TCF3 (E2A) fusion partner (in childhood Leukemia))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR