Anti-RNF8 Rabbit Polyclonal Antibody

Lieferant: Abgent
AP9820A
ABGEAP9820AEA 354 EUR
ABGEAP9820A
Anti-RNF8 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: RNF8 (ring finger protein 8, E3 ubiquitin protein ligase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR