Anti-SPR Rabbit Polyclonal Antibody

Lieferant: Abgent
AP9950B
ABGEAP9950BEA 354 EUR
ABGEAP9950B
Anti-SPR Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SPR (sepiapterin reductase (7,8-dihydrobiopterin:NADP+ oxidoreductase))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR