Anti-STEAP1 CHO Cells Monoclonal Antibody
Lieferant: Abgent
APR10758
ABGEAPR10758EA 354 EUR
ABGEAPR10758
Anti-STEAP1 CHO Cells Monoclonal Antibody
Antikörper
Anti-STEAP1 Reference Antibody (vandortuzumAb) on SDS-PAGE under reducing (R) condition. The gel was stained with Coomassie Blue. The purity of the protein is greater than 95%.
  • Cellular location - Endosome membrane {ECO:0000250|UniProtKB:Q9CWR7}; multi-pass membrane protein, cell membrane, multi-pass membrane protein
  • Tissue location - Ubiquitously expressed, highly expressed in prostate tumours
Does not function as a metalloreductase due to the absence of binding sites for the electron-donating substrate NADPH. Promotes Fe(3⁺) reduction when fused to the NADPH-binding domain of STEAP4.
Type: Primary
Antigen: STEAP1
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: CHO cells
Isotype: IgG1
Reactivity: Cynomolgus monkey, human