Anti-HOXD11A Rabbit Polyclonal Antibody

Lieferant: Abgent
AZB10017A
ABGEAZB10017AEA 354 EUR
ABGEAZB10017A
Anti-HOXD11A Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: HOXD11A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype:
Reactivity: Zebrafish
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR