Anti-TFAP2E Rabbit Polyclonal Antibody

Lieferant: Abgent
AZB10019A
ABGEAZB10019AEA 354 EUR
ABGEAZB10019A
Anti-TFAP2E Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TFAP2E
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype:
Reactivity: Zebrafish
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR