Anti-AHCY Mouse Monoclonal Antibody [clone: M1]

Lieferant: Abnova
H00000191-M08A
ABNOH00000191-M08AEA 397 EUR
ABNOH00000191-M08A
Anti-AHCY Mouse Monoclonal Antibody [clone: M1]
Antikörper
Type: Primary
Antigen: AHCY
Clonality: Monoclonal
Clone: M1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR