Anti-ABCC6 Mouse Monoclonal Antibody [clone: 5D8]

Lieferant: Abnova
H00000368-M02
ABNOH00000368-M02EA 397 EUR
ABNOH00000368-M02
Anti-ABCC6 Mouse Monoclonal Antibody [clone: 5D8]
Antikörper
Type: Primary
Antigen: ABCC6
Clonality: Monoclonal
Clone: 5D8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR