Anti-B2M Mouse Polyclonal Antibody

Lieferant: Abnova
H00000567-A01
ABNOH00000567-A01EA 260 EUR
ABNOH00000567-A01
Anti-B2M Mouse Polyclonal Antibody
Antikörper
Type: Primary
Antigen: B2M
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR