Anti-BMP2 Mouse Monoclonal Antibody [clone: 4B12]

Lieferant: Abnova
H00000650-M03
ABNOH00000650-M03EA 397 EUR
ABNOH00000650-M03
Anti-BMP2 Mouse Monoclonal Antibody [clone: 4B12]
Antikörper
Type: Primary
Antigen: BMP2
Clonality: Monoclonal
Clone: 4B12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR