Anti-C1QC Mouse Polyclonal Antibody
Lieferant: Abnova
H00000714-B01P
ABNOH00000714-B01PEA 397 EUR
ABNOH00000714-B01P
Anti-C1QC Mouse Polyclonal Antibody
Antikörper
Type: Primary
Antigen: C1QC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human