Anti-CAMK2G Mouse Polyclonal Antibody
Lieferant: Abnova
H00000818-B02P
ABNOH00000818-B02PEA 397 EUR
ABNOH00000818-B02P
Anti-CAMK2G Mouse Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CAMK2G
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Rat