Anti-CDK7 Mouse Polyclonal Antibody
Lieferant: Abnova
H00001022-B01P
ABNOH00001022-B01PEA 397 EUR
ABNOH00001022-B01P
Anti-CDK7 Mouse Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CDK7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human