Anti-COX6B1 Mouse Monoclonal Antibody [clone: 2D3]

Lieferant: Abnova
H00001340-M02
ABNOH00001340-M02EA 397 EUR
ABNOH00001340-M02
Anti-COX6B1 Mouse Monoclonal Antibody [clone: 2D3]
Antikörper
Type: Primary
Antigen: COX6B1
Clonality: Monoclonal
Clone: 2D3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR