Anti-CTLA4 Mouse Monoclonal Antibody (Biotin)

Lieferant: Abnova
H00001493-AP41
ABNOH00001493-AP41EA 842 EUR
ABNOH00001493-AP41
Anti-CTLA4 Mouse Monoclonal Antibody (Biotin)
Antikörper
This Set comes with matched Antibody Pair to detect and quantify protein level of human CTLA4.

Antibody pair set content: 1. Capture Antibody: Mouse Monoclonal Anti-CTLA4, IgG1 kappa (100 ?g); 2. Detection Antibody: biotinylated Mouse Monoclonal Anti-CTLA4, IgG1 kappa (50 ?g);

Type:
Antigen: CTLA4
Clonality: Monoclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR