Anti-CTSL Rabbit Polyclonal Antibody
Lieferant: Abnova
H00001514-D01P
ABNOH00001514-D01PEA 364 EUR
ABNOH00001514-D01P
Anti-CTSL Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CTSL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse