Anti-DLX2 Mouse Monoclonal Antibody [clone: 3H3]

Lieferant: Abnova
H00001746-M07
ABNOH00001746-M07EA 397 EUR
ABNOH00001746-M07
Anti-DLX2 Mouse Monoclonal Antibody [clone: 3H3]
Antikörper
Type: Primary
Antigen: DLX2
Clonality: Monoclonal
Clone: 3H3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR