Anti-ELF3 Mouse Monoclonal Antibody [clone: 2B11]
Lieferant: Abnova
H00001999-M02
ABNOH00001999-M02EA 397 EUR
ABNOH00001999-M02
Anti-ELF3 Mouse Monoclonal Antibody [clone: 2B11]
Antikörper
Type: Primary
Antigen: ELF3
Clonality: Monoclonal
Clone: 2B11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human