Anti-ELF3 Mouse Monoclonal Antibody [clone: 3B6]

Lieferant: Abnova
H00001999-M06
ABNOH00001999-M06EA 397 EUR
ABNOH00001999-M06
Anti-ELF3 Mouse Monoclonal Antibody [clone: 3B6]
Antikörper
Type: Primary
Antigen: ELF3
Clonality: Monoclonal
Clone: 3B6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR