Anti-ELF4 Mouse Monoclonal Antibody [clone: 1E10]

Lieferant: Abnova
H00002000-M02
ABNOH00002000-M02EA 397 EUR
ABNOH00002000-M02
Anti-ELF4 Mouse Monoclonal Antibody [clone: 1E10]
Antikörper
Type: Primary
Antigen: ELF4
Clonality: Monoclonal
Clone: 1E10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR