Anti-ELF5 Mouse Monoclonal Antibody [clone: 3D10]

Lieferant: Abnova
H00002001-M01
ABNOH00002001-M01EA 397 EUR
ABNOH00002001-M01
Anti-ELF5 Mouse Monoclonal Antibody [clone: 3D10]
Antikörper
Type: Primary
Antigen: ELF5
Clonality: Monoclonal
Clone: 3D10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR