Anti-MARK2 Mouse Monoclonal Antibody [clone: 3C5]
Lieferant: Abnova
H00002011-M02
ABNOH00002011-M02EA 397 EUR
ABNOH00002011-M02
Anti-MARK2 Mouse Monoclonal Antibody [clone: 3C5]
Antikörper
Type: Primary
Antigen: MARK2
Clonality: Monoclonal
Clone: 3C5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human