Anti-FKBP1A Mouse Monoclonal Antibody [clone: 1E5-A12]

Lieferant: Abnova
H00002280-M01
ABNOH00002280-M01EA 397 EUR
ABNOH00002280-M01
Anti-FKBP1A Mouse Monoclonal Antibody [clone: 1E5-A12]
Antikörper
Type: Primary
Antigen: FKBP1A
Clonality: Monoclonal
Clone: 1E5-A12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR