Anti-ACKR1 Rabbit Polyclonal Antibody
Lieferant: Abnova
H00002532-D01
ABNOH00002532-D01EA 364 EUR
ABNOH00002532-D01
Anti-ACKR1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ACKR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human