Anti-CXCL2 Mouse Monoclonal Antibody [clone: 2D10]

Lieferant: Abnova
H00002920-M21
ABNOH00002920-M21EA 397 EUR
ABNOH00002920-M21
Anti-CXCL2 Mouse Monoclonal Antibody [clone: 2D10]
Antikörper
Mouse monoclonal antibody raised against a partial recombinant CXCL2.

Recommended Dilutions: Western Blot: 1-5 ?g/ml

Type: Primary
Antigen: CXCL2
Clonality: Monoclonal
Clone: 2D10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR