Anti-GTF2E1 Polyclonal Antibody Pair

Lieferant: Abnova
H00002960-AP22
ABNOH00002960-AP22EA 842 EUR
ABNOH00002960-AP22
Anti-GTF2E1 Polyclonal Antibody Pair
Antikörper
This Set comes with matched Antibody Pair to detect and quantify protein level of human GTF2E1.

Antibody pair set content: 1. Capture Antibody: Rabbit MaxPab® affinity purified Polyclonal Anti-GTF2E1 (100 µg); 2. Detection Antibody: Mouse purified Polyclonal Anti-GTF2E1 (20 µg);

Type:
Antigen: GTF2E1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR