Anti-GTF2H4 Polyclonal Antibody Pair

Lieferant: Abnova
H00002968-PW1
ABNOH00002968-PW1EA 842 EUR
ABNOH00002968-PW1
Anti-GTF2H4 Polyclonal Antibody Pair
Antikörper
This Immunopreciptitation/ Western Blot Antibody Pair set comes with one Antibody for Immunoprecipitation and another to detect the precipitated protein in Western Blot.

Antibody pair set content: 1. Antibody pair for IP: Rabbit Polyclonal Anti-GTF2H4 (300 µl); 2. Antibody pair for Western Blot: Mouse purified Polyclonal Anti-GTF2H4 (50 µg)

Type:
Antigen: GTF2H4
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR