Anti-CFH Rabbit Polyclonal Antibody

Lieferant: Abnova
H00003075-D01P
ABNOH00003075-D01PEA 364 EUR
ABNOH00003075-D01P
Anti-CFH Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CFH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR