Anti-ICAM4 Rabbit Polyclonal Antibody
Lieferant: Abnova
H00003386-D01P
ABNOH00003386-D01PEA 364 EUR
ABNOH00003386-D01P
Anti-ICAM4 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ICAM4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human