Anti-IL13RA1 Mouse Polyclonal Antibody

Lieferant: Abnova
H00003597-B01P
ABNOH00003597-B01PEA 397 EUR
ABNOH00003597-B01P
Anti-IL13RA1 Mouse Polyclonal Antibody
Antikörper
Mouse polyclonal antibody raised against a full-length human IL13RA1 protein.

Recommended Dilutions: Western Blot (1 µg/ml)

Type: Primary
Antigen: IL13RA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR